Learn More
Medchemexpress LLC D-glucose 6-phosphate-13C6 disodium xhydrate | 00-00-0 | MFCD00136040 | 99.6% | 310.06 g·mol⁻¹ | 13C6H11Na2O9P | 5 MG
SDP
Catalog No. 5000262412
Encompass_Preferred
D-Glucose 6-phosphate-13C6 disodium xhydrate is the 13C6 isotopically labeled form of glucose 6-phosphate supplied as the disodium salt. It is intended for use as a tracer and as an internal standard for quantitative analyses by NMR, GC-MS, or LC-MS, and is supplied at high purity for analytical workflows.
  • 13C6 isotopic labeling for tracer and internal standard applications.
  • Supplied as the disodium salt for improved solubility and handling.
  • High purity (99.6%) suitable for quantitative analysis.
  • Available in small analytical package sizes for method development.
  • Stable when stored sealed away from moisture and light; in solvent, -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-62412 Supplier Medchemexpress LLC Supplier No. HY112537S15MG
D-Glucose 6-phosphate-13C6 disodium xhydrate is the 13C6 isotopically labeled form of glucose 6-phosphate supplied as the disodium salt. It is intended for use as a tracer and as an internal standard for quantitative analyses by NMR, GC-MS, or LC-MS, and is supplied at high purity for analytical workflows.
  • 13C6 isotopic labeling for tracer and internal standard applications.
  • Supplied as the disodium salt for improved solubility and handling.
  • High purity (99.6%) suitable for quantitative analysis.
  • Available in small analytical package sizes for method development.
  • Stable when stored sealed away from moisture and light; in solvent, -80°C (6 months) or -20°C (1 month).