Learn More
Medchemexpress LLC D-Glucose 6-Phosphate-13C6 disodium xhydrate | 56-73-5 | 99.6% | 310.06 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY112537S11MG
Encompass_Preferred
D-Glucose 6-Phosphate-13C6 disodium xhydrate is the 13C labeled D-Glucose 6-phosphate. D-Glucose 6-phosphate is a glucose sugar phosphorylated at the hydroxy group on carbon 6. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-36343