Learn More
Medchemexpress LLC D-Glucose 6-Phosphate-13C6 disodium xhydrate | 56-73-5 | 99.6% | 310.06 | 10 MG
SDP
Catalog No. 5000438157
Encompass_Preferred
D-Glucose 6-Phosphate-13C6 disodium xhydrate is the 13C labeled D-Glucose 6-phosphate. It is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This compound is used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Labeled with 13C isotope
  • Functions as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Utilizes stable heavy isotopes for drug development quantitation
Catalog No. 50-004-38157 Supplier Medchemexpress LLC Supplier No. HY112537S110MG
D-Glucose 6-Phosphate-13C6 disodium xhydrate is the 13C labeled D-Glucose 6-phosphate. It is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This compound is used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Labeled with 13C isotope
  • Functions as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Utilizes stable heavy isotopes for drug development quantitation