Learn More
Medchemexpress LLC D-Glucose 6-phosphate | 56-73-5 | 99.9% | 260.14 | 25 MG
SDP
Catalog No. 5000438925
Encompass_Preferred
D-Glucose 6-phosphate is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is very common in cells, as the majority of glucose entering a cell becomes phosphorylated in this manner. It is an endogenous metabolite.
  • Phosphorylated glucose sugar
  • Endogenous metabolite
  • Very common in cells
  • For research use only
Catalog No. 50-004-38925 Supplier Medchemexpress LLC Supplier No. 25MGHY112537
D-Glucose 6-phosphate is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is very common in cells, as the majority of glucose entering a cell becomes phosphorylated in this manner. It is an endogenous metabolite.
  • Phosphorylated glucose sugar
  • Endogenous metabolite
  • Very common in cells
  • For research use only