Learn More
Selleck Chemical LLC D-Glucose 6-phosphate disodium
SDP
Catalog No. 5000003524
Encompass_Preferred
D-Glucose 6-phosphate disodium salt is the disodium salt form of D-Glucose 6-phosphate a common form of glucose within the cell participating in two major metabolic pathways glycolysis and the pentose phosphate pathway It can be coverted to glycogen or starch for storage
Catalog No. 50-000-03524 Supplier Selleck Chemical LLC Supplier No. S55181G
D-Glucose 6-phosphate disodium salt is the disodium salt form of D-Glucose 6-phosphate a common form of glucose within the cell participating in two major metabolic pathways glycolysis and the pentose phosphate pathway It can be coverted to glycogen or starch for storage