Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-glucose 6-phosphate disodium salt | 3671-99-6 | 99.9% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY128374R10MG

Encompass_Preferred

D-Glucose 6-phosphate disodium salt is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is very common in cells, as the majority of glucose entering a cell will become phosphorylated in this way, playing a crucial role as an endogenous metabolite.

  • Purity of 99.93%
  • Molecular formula: C6H11Na2O9P
  • Molecular weight: 304.10
  • Appears as a white to off-white solid
  • Highly soluble in water and PBS
  • Recommended storage for solid form: 4°C, sealed, away from moisture
  • Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month
  • Classified under ketones, aldehydes, and acids

Catalog No. 50-003-38918


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.