Learn More
Medchemexpress LLC D-Glucose 6-phosphate disodium salt | 3671-99-6 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY128374R50MG
Encompass_Preferred
D-Glucose 6-phosphate disodium salt is an endogenous metabolite, a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is commonly found in cells, as most glucose entering a cell undergoes this phosphorylation. It is intended for research use only.
  • High purity of 99.9%
  • Solid, white to off-white appearance
  • Ships at room temperature in continental US
  • Store at 4°C, sealed, away from moisture (in solvent: -80°C for 6 months; -20°C for 1 month)
  • Highly soluble in water (233.33 mg/mL) and PBS (16.67 mg/mL)
  • Functions as an endogenous metabolite and potential inhibitor
Catalog No. 50-003-39992