Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucose 6-phosphate disodium salt | 3671-99-6 | 99.9% | 500 MG
SDP

Catalog No. 5000339248
Encompass_Preferred

D-Glucose-6-phosphate disodium salt is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is very common in cells, as the majority of glucose entering a cell will become phosphorylated in this way.

  • White to off-white solid appearance
  • Molecular weight: 304.10
  • Initial source: Endogenous metabolite
  • Can be stored at 4°C, sealed, away from moisture
  • Soluble in water for in vitro and in vivo applications

Catalog No. 50-003-39248 Supplier Medchemexpress LLC Supplier No. HY128374500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucose-6-phosphate disodium salt is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is very common in cells, as the majority of glucose entering a cell will become phosphorylated in this way.

  • White to off-white solid appearance
  • Molecular weight: 304.10
  • Initial source: Endogenous metabolite
  • Can be stored at 4°C, sealed, away from moisture
  • Soluble in water for in vitro and in vivo applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.