Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucose 6-Phosphate Disodium Salt Dihydrate | 3671-99-6 | 99.9% | 25MG
SDP

Catalog No. 5000342405
Encompass_Preferred

D-Glucose-6-phosphate disodium salt is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is very common in cells, as most glucose entering a cell becomes phosphorylated in this manner.

  • Purity: 99.9%
  • Molecular weight: 304.10
  • Formula: C6H11Na2O9P
  • Appearance: Solid, white to off-white
  • Soluble in water and PBS
  • Classified as a ketone, aldehyde, and acid
  • Endogenous metabolite

Catalog No. 50-003-42405 Supplier Medchemexpress LLC Supplier No. 25MGHY128374R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucose-6-phosphate disodium salt is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is very common in cells, as most glucose entering a cell becomes phosphorylated in this manner.

  • Purity: 99.9%
  • Molecular weight: 304.10
  • Formula: C6H11Na2O9P
  • Appearance: Solid, white to off-white
  • Soluble in water and PBS
  • Classified as a ketone, aldehyde, and acid
  • Endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.