Learn More
TARGETMOL CHEMICALS INC D-GLUCOSE 6-PHOSPHATE SO 50MG
SDP
Catalog No. 502576732
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg and bulk. Please contact Fisher for quotes. In cells D-glucose 6-phosphate (G6P) is generated when glucose is phosphorylated by hexokinase or glucokinase or by the conversion of glucose-1-phosphate by phosphoglucomutase during glycogenolysis. G6P lies at the beginning of both glycolysis and the pentose phosphate pathways. It also can be stored as glycogen when blood glucose levels are high. purity: 100%
Catalog No. 50-257-6732 Supplier TARGETMOL CHEMICALS INC Supplier No. T472650MG
Also available in 5 mg 10 mg 25 mg 100 mg and bulk. Please contact Fisher for quotes. In cells D-glucose 6-phosphate (G6P) is generated when glucose is phosphorylated by hexokinase or glucokinase or by the conversion of glucose-1-phosphate by phosphoglucomutase during glycogenolysis. G6P lies at the beginning of both glycolysis and the pentose phosphate pathways. It also can be stored as glycogen when blood glucose levels are high. purity: 100%