Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucose 6-phosphate sodium | 54010-71-8 | 98.0% | 282.12 | 100 MG
SDP

Catalog No. 5000438156
Encompass_Preferred

D-Glucose 6-phosphate sodium is an endogenous metabolite for research use only. It is not sold to patients.

  • Human endogenous metabolite
  • Appears as a white to off-white solid
  • Store at 4°C under nitrogen
  • Soluble in H2O at 50 mg/mL (needs ultrasonic)
  • Classified as a ketone, aldehyde, and acid

Catalog No. 50-004-38156 Supplier Medchemexpress LLC Supplier No. HY112537B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucose 6-phosphate sodium is an endogenous metabolite for research use only. It is not sold to patients.

  • Human endogenous metabolite
  • Appears as a white to off-white solid
  • Store at 4°C under nitrogen
  • Soluble in H2O at 50 mg/mL (needs ultrasonic)
  • Classified as a ketone, aldehyde, and acid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.