Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucose 6-phosphate sodium | 54010-71-8 | 98.0% | 282.12 | 5 G
SDP

Catalog No. 5000436325
Encompass_Preferred

D-Glucose 6-phosphate sodium is an endogenous metabolite used for research purposes. It is not sold to patients.

  • Endogenous metabolite
  • For research use only
  • Human endogenous metabolite
  • Solid appearance
  • White to off-white color
  • Soluble in H2O (50 mg/mL)
  • Store at 4°C under nitrogen

Catalog No. 50-004-36325 Supplier Medchemexpress LLC Supplier No. HY112537B5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucose 6-phosphate sodium is an endogenous metabolite used for research purposes. It is not sold to patients.

  • Endogenous metabolite
  • For research use only
  • Human endogenous metabolite
  • Solid appearance
  • White to off-white color
  • Soluble in H2O (50 mg/mL)
  • Store at 4°C under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.