Learn More
Medchemexpress LLC D-Glucose 6-phosphate sodium | 54010-71-8 | 98.0% | 282.12 | 500 MG

Supplier: Medchemexpress LLC HY112537B500MG
D-Glucose 6-phosphate sodium is an endogenous metabolite. It is for research use only and serves as a human endogenous metabolite.
- Solid appearance.
- White to off-white color.
- Shipping at room temperature in continental US; may vary elsewhere.
- Storage at 4°C, stored under nitrogen.
- In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen).
- Soluble in H2O: 50 mg/mL (177.23 mM; requires ultrasonic).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.