Learn More
Cayman Chemical D-Glucose-6-phosphate (sodium salt), Purity: ≥98%, CAS: 54010-71-8, 5 g
SDP
Catalog No. 502984795
Encompass_Preferred
A fundamental component of glucose metabolism involved in the homeostatic regulation of blood glucose levels by participating in glycolysis, glycogen synthesis, and gluconeogenesis
Catalog No. 50-298-4795 Supplier Cayman Chemical Supplier No. 203765 G
A fundamental component of glucose metabolism involved in the homeostatic regulation of blood glucose levels by participating in glycolysis, glycogen synthesis, and gluconeogenesis