Learn More
Research Products International Corp D-(+)-Glucose [Dextrose Anhydrous], ACS Grade, 500 Grams

Supplier: Research Products International Corp G32045500.0
Substance Name: Dextrose Anhydrous
CAS Number: 50-99-7
Molecular Formula: C6H12O6
Molecular Weight: 180.2
Chemical Purity: >99.8%
Soluble in:?Water
Store at:?Room Temperature
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.