Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucose, Anhydrous, Granular, ACS, Spectrum™ Chemical
SDP

Catalog No. 18610160 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-160 100 lb. Fiber Drum
18-610-161 2.5 kg Poly Bottle
18-610-162 12 kg Poly Pail
18-610-163 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-610-160 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1017100LBBL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
IUPAC Name 6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OCC1OC(O)C(O)C(O)C1O

Specifications

CAS Max % 1
Ignition Residue 0.0002
Loss on Drying 0.002
Packaging Fiber Drum
Quantity 100 lb.
Specific Rotation +52.5° to +53.0°
Grade ACS
Insoluble Matter 5e-05
Titratable Acid 0.002 meq/g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.