Learn More
Sigma Aldrich Fine Chemicals Biosciences D Glucose BioXtra 99.5 G5KG
Catalog No. 502902144
Encompass
Glucose/dextrose is a carbohydrate compound or an aldohexose present in humans. It is used as fuel by cells. Glucose consists of six carbon atoms and an aldehyde group. This monosaccharide can exist in an open-chain (acyclic) and ring (cyclic) form. It plays an important role in animal metabolismGlucose a major product of photosynthesis participates in cellular respiration to generate adenosine triphosphate (ATP) and NADH (nicotinamide adenine dinucleotide).
Catalog No. 50-290-2144 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G75285KG
Glucose/dextrose is a carbohydrate compound or an aldohexose present in humans. It is used as fuel by cells. Glucose consists of six carbon atoms and an aldehyde group. This monosaccharide can exist in an open-chain (acyclic) and ring (cyclic) form. It plays an important role in animal metabolismGlucose a major product of photosynthesis participates in cellular respiration to generate adenosine triphosphate (ATP) and NADH (nicotinamide adenine dinucleotide).