Learn More
Medchemexpress LLC D-(+)-glucose-d1-3 | 106032-60-4 | 99.9% | 181.16 g/mol | C6H11DO6 | 10 MG

Supplier: Medchemexpress LLC HYB0389S710MG
D-Glucose-d1-3 is a deuterium-labeled form of D-(+)-glucose with a deuterium atom substituted at positions 1-3. It is provided as a high-purity labeled standard for tracer studies, metabolic research, and analytical applications.
- Deuterium-labeled at positions 1-3.
- Cas number 106032-60-4.
- Molecular formula C6H11DO6.
- Molecular weight 181.16 g/mol.
- Purity 99.9%.
- Available as a 10 mg vial.
- Storage: powder -20°C for long-term; in solvent -80°C for up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.