Learn More
Medchemexpress LLC D-Glucose-d1-3 | 106032-60-4 | 99.9% | 5 MG
SDP
Catalog No. 5000405422
Encompass_Preferred
D-Glucose-d1-3 is the deuterium labeled D-Glucose. D-Glucose, a monosaccharide, is an important carbohydrate and critical component of general metabolism and signaling pathways.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-05422 Supplier Medchemexpress LLC Supplier No. HYB0389S75MG
Add to Cart
Add to Cart
D-Glucose-d1-3 is the deuterium labeled D-Glucose. D-Glucose, a monosaccharide, is an important carbohydrate and critical component of general metabolism and signaling pathways.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.