Learn More
Medchemexpress LLC D-Glucose-d1-4 | 106032-61-5 | 98.0% | 100 MG
SDP
Catalog No. 5000407128
Encompass_Preferred
D-Glucose-d1-4 is the deuterium labeled D-Glucose. This monosaccharide is an important carbohydrate in biology, serving as a critical component of general metabolism and a significant signaling molecule for cellular metabolic status and stress response.
  • Deuterium labeled D-Glucose
  • Important carbohydrate in biology
  • Critical component of general metabolism
  • Significant signaling molecule
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07128 Supplier Medchemexpress LLC Supplier No. HYB0389S8100MG
D-Glucose-d1-4 is the deuterium labeled D-Glucose. This monosaccharide is an important carbohydrate in biology, serving as a critical component of general metabolism and a significant signaling molecule for cellular metabolic status and stress response.
  • Deuterium labeled D-Glucose
  • Important carbohydrate in biology
  • Critical component of general metabolism
  • Significant signaling molecule
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS