Learn More
Medchemexpress LLC D-Glucose-d1-4 | 106032-61-5 | 98.0% | 50 MG
SDP
Catalog No. 5000407147
Encompass_Preferred
D-Glucose-d1-4 is the deuterium labeled D-Glucose, a monosaccharide and important carbohydrate in biology. It serves as a carbohydrate sweetener and plays critical roles in general metabolism, cellular metabolic status, and stress response. This compound is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen and carbon
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-07147 Supplier Medchemexpress LLC Supplier No. HYB0389S850MG
D-Glucose-d1-4 is the deuterium labeled D-Glucose, a monosaccharide and important carbohydrate in biology. It serves as a carbohydrate sweetener and plays critical roles in general metabolism, cellular metabolic status, and stress response. This compound is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen and carbon
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs