Learn More
Medchemexpress LLC D-Glucose-d1 | 56570-89-9 | 99.9% | 1 MG
SDP
Catalog No. 5000407144
Encompass_Preferred
D-Glucose-d is a deuterium labeled D-Glucose, which is an important monosaccharide and carbohydrate in biology. It serves as a critical component of general metabolism and acts as a signaling molecule. This compound is suitable for various quantitative analyses and can impact pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-07144 Supplier Medchemexpress LLC Supplier No. HYB0389S11MG
D-Glucose-d is a deuterium labeled D-Glucose, which is an important monosaccharide and carbohydrate in biology. It serves as a critical component of general metabolism and acts as a signaling molecule. This compound is suitable for various quantitative analyses and can impact pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution may affect pharmacokinetic and metabolic profiles of drugs