Learn More
Medchemexpress LLC D-Glucose-d12-1 | 89798-27-6 | 99.4% | 100 MG
SDP
Catalog No. 5000407267
Encompass_Preferred
D-Glucose-d12-1 is a deuterium-labeled D-Glucose, which is a monosaccharide essential for general metabolism and acts as a critical signaling molecule in cellular metabolic status and stress response. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07267 Supplier Medchemexpress LLC Supplier No. HYB0389S2100MG
D-Glucose-d12-1 is a deuterium-labeled D-Glucose, which is a monosaccharide essential for general metabolism and acts as a critical signaling molecule in cellular metabolic status and stress response. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS