Learn More
Medchemexpress LLC D-(+)-glucose-d12-1 | 89798-27-6 | 99.4% | 192.23 g/mol | C6D12O6 | 10 MG
SDP
Catalog No. 5000298060
Encompass_Preferred
D-Glucose-d12-1 is a deuterium-labeled form of D-glucose (C6D12O6) intended for use as an isotopic tracer and as a quantitative internal standard in analytical methods. It is supplied as a high-purity crystalline powder for laboratory research and analytical workflows.
  • Uniformly deuterated glucose (C6D12O6).
  • High purity: 99.4%.
  • Used as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Supplied as a stable crystalline powder suitable for analytical preparation.
  • Recommended storage: powder at -20°C for long-term stability.
Catalog No. 50-002-98060 Supplier Medchemexpress LLC Supplier No. HYB0389S210MG
D-Glucose-d12-1 is a deuterium-labeled form of D-glucose (C6D12O6) intended for use as an isotopic tracer and as a quantitative internal standard in analytical methods. It is supplied as a high-purity crystalline powder for laboratory research and analytical workflows.
  • Uniformly deuterated glucose (C6D12O6).
  • High purity: 99.4%.
  • Used as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Supplied as a stable crystalline powder suitable for analytical preparation.
  • Recommended storage: powder at -20°C for long-term stability.