Learn More
Medchemexpress LLC D-Glucose-d12-1 | 89798-27-6 | 99.4% | 5 MG
SDP
Catalog No. 5000406408
Encompass_Preferred
D-Glucose-d12-1 is the deuterium labeled D-Glucose. D-Glucose (Glucose), a monosaccharide, is an important carbohydrate in biology. It is a carbohydrate sweetener and a critical component of general metabolism, serving as a critical signaling molecule in relation to both cellular metabolic status and biotic and abiotic stress response.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06408 Supplier Medchemexpress LLC Supplier No. HYB0389S25MG
D-Glucose-d12-1 is the deuterium labeled D-Glucose. D-Glucose (Glucose), a monosaccharide, is an important carbohydrate in biology. It is a carbohydrate sweetener and a critical component of general metabolism, serving as a critical signaling molecule in relation to both cellular metabolic status and biotic and abiotic stress response.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.