Learn More
Medchemexpress LLC D-Glucose-d12-1 | 89798-27-6 | 99.4% | 50 MG
SDP
Catalog No. 5000407321
Encompass_Preferred
D-Glucose-d12-1 is a deuterium-labeled D-Glucose. It is a monosaccharide, a crucial carbohydrate in biology, a carbohydrate sweetener, and a vital component of general metabolism. It also serves as a critical signaling molecule related to cellular metabolic status and biotic and abiotic stress responses.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
  • Deuteration can potentially affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07321 Supplier Medchemexpress LLC Supplier No. HYB0389S250MG
D-Glucose-d12-1 is a deuterium-labeled D-Glucose. It is a monosaccharide, a crucial carbohydrate in biology, a carbohydrate sweetener, and a vital component of general metabolism. It also serves as a critical signaling molecule related to cellular metabolic status and biotic and abiotic stress responses.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
  • Deuteration can potentially affect pharmacokinetic and metabolic profiles of drugs.