Learn More
Medchemexpress LLC D-Glucose-d2 | 18991-62-3 | MFCD00064367 | 99.9% | 50 MG
SDP
Catalog No. 5000406506
Encompass_Preferred
D-Glucose-d2 is a deuterium-labeled form of D-Glucose. D-Glucose, a monosaccharide, plays a vital role in biology as an important carbohydrate. It acts as a carbohydrate sweetener, a critical component of general metabolism, and a signaling molecule related to cellular metabolic status and biotic and abiotic stress responses.
  • Can be utilized as a tracer.
  • Can serve as an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development processes.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06506 Supplier Medchemexpress LLC Supplier No. HYB0389S550MG
D-Glucose-d2 is a deuterium-labeled form of D-Glucose. D-Glucose, a monosaccharide, plays a vital role in biology as an important carbohydrate. It acts as a carbohydrate sweetener, a critical component of general metabolism, and a signaling molecule related to cellular metabolic status and biotic and abiotic stress responses.
  • Can be utilized as a tracer.
  • Can serve as an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development processes.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.