Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-glucose-d4 | 478529-49-6 | 99.0% | 184.18 g/mol | C6H8D4O6 | 10 MG
SDP

Catalog No. 5000298092
Encompass_Preferred

D-Glucose-d4 is a deuterium-labeled D-glucose used as an isotopic internal standard and tracer for analytical and metabolic studies. It is commonly applied in LC-MS, GC-MS, NMR, and metabolic flux experiments where stable isotope labeling enables tracking or accurate quantification.

  • Deuterium labeling: d4
  • Purity: 99.0%
  • Chemical formula: C6H8D4O6
  • Molecular weight: 184.18 g/mol
  • Appearance: solid, white to off-white
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Pack size: 10 MG

Catalog No. 50-002-98092 Supplier Medchemexpress LLC Supplier No. HYB0389S2410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucose-d4 is a deuterium-labeled D-glucose used as an isotopic internal standard and tracer for analytical and metabolic studies. It is commonly applied in LC-MS, GC-MS, NMR, and metabolic flux experiments where stable isotope labeling enables tracking or accurate quantification.

  • Deuterium labeling: d4
  • Purity: 99.0%
  • Chemical formula: C6H8D4O6
  • Molecular weight: 184.18 g/mol
  • Appearance: solid, white to off-white
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Pack size: 10 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.