Learn More
Medchemexpress LLC D-Glucose-d4 | 478529-49-6 | 99.0% | 50 MG
SDP
Catalog No. 5000405423
Encompass_Preferred
D-Glucose-d4 is the deuterium-labeled form of D-Glucose, an important monosaccharide and carbohydrate in biology. It serves as a critical signaling molecule and component of general metabolism. This compound can be utilized as a tracer and as an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-05423 Supplier Medchemexpress LLC Supplier No. HYB0389S2450MG
D-Glucose-d4 is the deuterium-labeled form of D-Glucose, an important monosaccharide and carbohydrate in biology. It serves as a critical signaling molecule and component of general metabolism. This compound can be utilized as a tracer and as an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.