Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-glucose-d7 | 66034-51-3 | 99.4% | 187.20 g·mol⁻¹ | C6H5D7O6 | 10 MG
SDP

Catalog No. 5000298061
Encompass_Preferred

D-Glucose-d7 is a deuterium-labeled form of D-glucose used as a stable isotope internal standard and tracer in analytical and metabolic studies. It is supplied as a white to off-white solid with high isotopic enrichment and high purity, suitable for mass spectrometry, metabolic flux analysis, and tracer experiments. Store as a powder at low temperatures for long-term stability.

  • Deuterium-labeled glucose for internal standard and tracer applications.
  • High isotopic enrichment (98.2%) and high purity (99.44%).
  • White to off-white solid suitable for analytical workflows.
  • Molecular formula C6H5D7O6; molecular weight 187.20 g·mol⁻¹.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-98061 Supplier Medchemexpress LLC Supplier No. HYB0389S610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucose-d7 is a deuterium-labeled form of D-glucose used as a stable isotope internal standard and tracer in analytical and metabolic studies. It is supplied as a white to off-white solid with high isotopic enrichment and high purity, suitable for mass spectrometry, metabolic flux analysis, and tracer experiments. Store as a powder at low temperatures for long-term stability.

  • Deuterium-labeled glucose for internal standard and tracer applications.
  • High isotopic enrichment (98.2%) and high purity (99.44%).
  • White to off-white solid suitable for analytical workflows.
  • Molecular formula C6H5D7O6; molecular weight 187.20 g·mol⁻¹.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.