Learn More
Alkali Scientific D (+) Glucose [Dextrose Anhydrous], ACS Grade, 10 Kg
SDP
Catalog No. 5000660975
Encompass_Preferred
D (+) Glucose Anhydrous (ACS Grade), 10 kg, is an analytical reagent used in various high-precision laboratory applications. This glucose is a primary energy source utilized in cellular respiration studies, fermentation experiments, and microbial culture media preparation. The ACS-grade ensures the highest level of purity, making it suitable for critical applications in pharmaceuticals, diagnostics, and biochemical research. The anhydrous form is preferred in situations where moisture content must be minimized, ensuring the reagent remains stable and accurate in chemical reactions. This large quantity is ideal for industrial and research laboratories that require high-quality glucose on a regular basis.
Catalog No. 50-006-60975 Supplier Alkali Scientific Supplier No. OSE001CP
D (+) Glucose Anhydrous (ACS Grade), 10 kg, is an analytical reagent used in various high-precision laboratory applications. This glucose is a primary energy source utilized in cellular respiration studies, fermentation experiments, and microbial culture media preparation. The ACS-grade ensures the highest level of purity, making it suitable for critical applications in pharmaceuticals, diagnostics, and biochemical research. The anhydrous form is preferred in situations where moisture content must be minimized, ensuring the reagent remains stable and accurate in chemical reactions. This large quantity is ideal for industrial and research laboratories that require high-quality glucose on a regular basis.