Learn More
Alkali Scientific D (+) Glucose [Dextrose Anhydrous], USP Grade, 1 Kg
SDP
Catalog No. 5000660836
Encompass_Preferred
D (+) Glucose Anhydrous (USP Grade), 1 kg, is a high-purity reagent used in various laboratory, pharmaceutical, and industrial applications. Glucose is a critical carbohydrate that serves as a primary energy source in both plant and animal cells. This anhydrous form of glucose is especially useful in situations where water content must be minimized, such as in dry formulations and precise chemical synthesis. In laboratory experiments, it is used in microbial culture media, fermentation processes, and glucose metabolism studies. The USP grade ensures the highest purity, making it suitable for pharmaceutical applications and quality control in scientific research.
Catalog No. 50-006-60836 Supplier Alkali Scientific Supplier No. COS001CP
D (+) Glucose Anhydrous (USP Grade), 1 kg, is a high-purity reagent used in various laboratory, pharmaceutical, and industrial applications. Glucose is a critical carbohydrate that serves as a primary energy source in both plant and animal cells. This anhydrous form of glucose is especially useful in situations where water content must be minimized, such as in dry formulations and precise chemical synthesis. In laboratory experiments, it is used in microbial culture media, fermentation processes, and glucose metabolism studies. The USP grade ensures the highest purity, making it suitable for pharmaceutical applications and quality control in scientific research.