Learn More
Alkali Scientific D (+) Glucose [Dextrose Anhydrous], USP Grade, 10 Kg
SDP
Catalog No. 5000660786
Encompass_Preferred
D (+) Glucose Anhydrous (USP Grade), 10 kg, is a bulk reagent used in industrial, laboratory, and pharmaceutical applications. This high-purity glucose is used extensively in research and production processes that require large quantities of glucose. It serves as a primary energy source in various biotechnological and biochemical processes, including fermentation, culture media preparation, and enzymatic reactions. The anhydrous form is ideal for applications where moisture must be minimized, ensuring stable storage and accurate formulation. This product complies with USP standards, ensuring the highest quality and purity for use in pharmaceutical formulations and laboratory experiments.
Catalog No. 50-006-60786 Supplier Alkali Scientific Supplier No. COS002CP
D (+) Glucose Anhydrous (USP Grade), 10 kg, is a bulk reagent used in industrial, laboratory, and pharmaceutical applications. This high-purity glucose is used extensively in research and production processes that require large quantities of glucose. It serves as a primary energy source in various biotechnological and biochemical processes, including fermentation, culture media preparation, and enzymatic reactions. The anhydrous form is ideal for applications where moisture must be minimized, ensuring stable storage and accurate formulation. This product complies with USP standards, ensuring the highest quality and purity for use in pharmaceutical formulations and laboratory experiments.