Learn More
Alkali Scientific D (+) Glucose [Dextrose Anhydrous], USP Grade, 5 Kg
SDP
Catalog No. 5000660769
Encompass_Preferred
D (+) Glucose Anhydrous (USP Grade), 5 kg, is a high-purity carbohydrate reagent used for large-scale applications in laboratories, pharmaceutical manufacturing, and industrial processes. This glucose is commonly utilized in research involving cellular energy metabolism, fermentation, and microbial culture media. The anhydrous form minimizes water content, making it ideal for precise formulations and experiments where moisture can interfere with reactions. This 5 kg quantity is designed for high-volume applications in pharmaceutical production, industrial processes, and large research projects that require consistent, high-quality glucose.
Catalog No. 50-006-60769 Supplier Alkali Scientific Supplier No. COS005CP
D (+) Glucose Anhydrous (USP Grade), 5 kg, is a high-purity carbohydrate reagent used for large-scale applications in laboratories, pharmaceutical manufacturing, and industrial processes. This glucose is commonly utilized in research involving cellular energy metabolism, fermentation, and microbial culture media. The anhydrous form minimizes water content, making it ideal for precise formulations and experiments where moisture can interfere with reactions. This 5 kg quantity is designed for high-volume applications in pharmaceutical production, industrial processes, and large research projects that require consistent, high-quality glucose.