Learn More
Bioworld D-Glucose, Monohydrate, 2.5 kg
Supplier:  Bioworld 407000122
Encompass_Preferred
Awhite granular monosaccharide.Obtained by enzymatic hydrolysis of starch to dextrose and subsequent purification and crystallization from solution.Sweet in taste.Specification:Biotechnology GradeSpecific rotation: +53.6 to +53.2°Insolubles: <0.0005%Residue after Ignition: <0.02%Chloride: <0.01%Sulfate and Sulfite: <0.005%Heavy Metals (as Pb): <0.0005%Iron: <0.0005%Moisture (105°C): 8-9%RNase and DNase, Protease activity: None detectedAppearance: SolidBoiling Point : 410.8 °CIUPAC_Name: (2s,3r,4r,5s,6r)-6-(Hydroxymethyl)Oxane-2,3,4,5-Tetrol HydrateMelting Point: 83 °C
Catalog No. 50-256-021