Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ D(+)-Glucose, Monohydrate, Calbiochem™,

Catalog No. 40744 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-744 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-744 Supplier MilliporeSigma™ Supplier No. 40744

missing translation for 'validMainframeMsg'

Suitable for biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14431-43-7
Molecular Formula C6H12O6·H2O

Specifications

Quantity 5 kg
Formula Weight 198.17
Chemical Name or Material D(+)-Glucose, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.