Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Glucose, MP Biomedicals™

Catalog No. MP21946721 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21946721 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21946721 Supplier MP Biomedicals, Inc Supplier No. 02194672.1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(+)-Glucose Anhydrous, Cell Culture Reagent, White Powder

  • D-(+)-Glucose, anhydrous is a main source of energy for living organisms.
  • It is combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen.
  • D-(+)-Glucose is a monosaccharide. It is a mixture of α- and β-anomers, primarily the α-anomer.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
Synonym Dextrose, Cartose, Cerelose, Corn Sugar

Specifications

Melting Point 150°C to 170°C (literature)
Color White
Density 1.544g/mL
Quantity 100 g
Beilstein 1724615
Solubility Information Soluble in 20% aqueous clear colorless solution.
Optical Rotation (c=10, Water, NH4OH) - +52.5 ± 5°
Grade Cell Culture Reagent
Physical Form Crystalline Powder
RTECSNumber LZ6600000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.