Learn More
Medchemexpress LLC D-Glucose, O-α-D-glucopyranosyl-(1→6)-O-α-D-glucopyranosyl-(1→4)- | 33401-87-5 | 99.95% | 504.44 | 25 MG
SDP
Catalog No. 5000439560
Encompass_Preferred
D-Panose is an oligosaccharide composed of glucose units linked by specific glycosidic bonds. It is a PAN-type oligosaccharide and a food ingredient based on isomaltooligosaccharides (IMOs). It can be detected with high sensitivity in SALDI-MS using magnesium oxide nanoparticles as the matrix.
  • Composed of glucose units.
  • PAN-type oligosaccharide.
  • Food ingredient based on isomaltooligosaccharides (IMOs).
  • High sensitivity detection in SALDI-MS.
  • For research use only.
Catalog No. 50-004-39560 Supplier Medchemexpress LLC Supplier No. 25MGHY111951
D-Panose is an oligosaccharide composed of glucose units linked by specific glycosidic bonds. It is a PAN-type oligosaccharide and a food ingredient based on isomaltooligosaccharides (IMOs). It can be detected with high sensitivity in SALDI-MS using magnesium oxide nanoparticles as the matrix.
  • Composed of glucose units.
  • PAN-type oligosaccharide.
  • Food ingredient based on isomaltooligosaccharides (IMOs).
  • High sensitivity detection in SALDI-MS.
  • For research use only.