Learn More
Medchemexpress LLC β-D-Glucose pentaacetate | 604-69-3 | 98.0% | 390.34 | 1 ML
SDP
Catalog No. 5000400821
Encompass_Preferred
β-D-Glucose pentaacetate (Penta-O-acetyl-β-D-glucopyranose) is a glucose derivative primarily utilized to investigate the mechanism of insulin secretion. It is for research use only.
  • Used to explore the mechanism of insulin secretion.
  • Available in various forms including solid, solution, and solid with solvent.
Catalog No. 50-004-00821 Supplier Medchemexpress LLC Supplier No. HY2230610MM/1ML
β-D-Glucose pentaacetate (Penta-O-acetyl-β-D-glucopyranose) is a glucose derivative primarily utilized to investigate the mechanism of insulin secretion. It is for research use only.
  • Used to explore the mechanism of insulin secretion.
  • Available in various forms including solid, solution, and solid with solvent.