Learn More
Medchemexpress LLC β-D-Glucose pentaacetate | 604-69-3 | 98.0% | 390.34 | 500 G
SDP
Catalog No. 5000401582
Encompass_Preferred
β-D-Glucose Pentaacetate is a glucose derivative primarily used to explore the mechanism of insulin secretion. It is provided as a white to off-white solid with high purity.
  • Provided as a white to off-white solid
  • Primarily used to investigate insulin secretion mechanism
  • High purity of 98.0%
  • Should be stored at 4°C, protected from light and under nitrogen
Catalog No. 50-004-01582 Supplier Medchemexpress LLC Supplier No. HY22306500G
β-D-Glucose Pentaacetate is a glucose derivative primarily used to explore the mechanism of insulin secretion. It is provided as a white to off-white solid with high purity.
  • Provided as a white to off-white solid
  • Primarily used to investigate insulin secretion mechanism
  • High purity of 98.0%
  • Should be stored at 4°C, protected from light and under nitrogen