Learn More
Sigma Aldrich Fine Chemicals Biosciences D- -Glucose powder BioReag
Catalog No. 502902160
Encompass
Glucose is the energy source of brain and of cells containing few or no mitochondria. It is also a substrate for anaerobic glycolysis. The human body stores glucose in the form of glycogen. Under low glucose conditions it is released from liver and kidney via glycogen. Glucose is also required for the formation of di- oligo- and polysaccharides.
Catalog No. 50-290-2160 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G702110KG
Glucose is the energy source of brain and of cells containing few or no mitochondria. It is also a substrate for anaerobic glycolysis. The human body stores glucose in the form of glycogen. Under low glucose conditions it is released from liver and kidney via glycogen. Glucose is also required for the formation of di- oligo- and polysaccharides.