Learn More
Medchemexpress LLC D-Glucose (Standard) | 50-99-7 | 99.8% | 100 MG
SDP
Catalog No. 5000405421
Encompass_Preferred
D-Glucose (Standard) is the analytical standard of D-Glucose. It is the naturally occurring form of glucose and the most abundant monosaccharide, playing a critical role in general metabolism and cellular metabolic status signaling. This product is intended for research and analytical applications.
  • Analytical standard for D-Glucose
  • Naturally occurring form of glucose
  • Most abundant monosaccharide
  • Critical component of general metabolism
  • Significant signaling molecule for cellular metabolic status and stress response
  • Used as a reference standard for assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-05421 Supplier Medchemexpress LLC Supplier No. HYB0389R100MG
D-Glucose (Standard) is the analytical standard of D-Glucose. It is the naturally occurring form of glucose and the most abundant monosaccharide, playing a critical role in general metabolism and cellular metabolic status signaling. This product is intended for research and analytical applications.
  • Analytical standard for D-Glucose
  • Naturally occurring form of glucose
  • Most abundant monosaccharide
  • Critical component of general metabolism
  • Significant signaling molecule for cellular metabolic status and stress response
  • Used as a reference standard for assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS