Learn More
Medchemexpress LLC D-Glucose (Standard) | 50-99-7 | 99.8% | 250 MG
SDP
Catalog No. 5000407048
Encompass_Preferred
D-Glucose (Standard) is the analytical standard of D-Glucose, the naturally occurring and most abundant monosaccharide. It is intended for research and analytical applications, serving as a critical component of general metabolism and a signaling molecule in cellular metabolic status and stress response.
  • Analytical standard grade
  • Used as a reference standard assay
  • Commonly used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-004-07048 Supplier Medchemexpress LLC Supplier No. 250MGHYB0389R
D-Glucose (Standard) is the analytical standard of D-Glucose, the naturally occurring and most abundant monosaccharide. It is intended for research and analytical applications, serving as a critical component of general metabolism and a signaling molecule in cellular metabolic status and stress response.
  • Analytical standard grade
  • Used as a reference standard assay
  • Commonly used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)