Learn More
Medchemexpress LLC D-Glucose (Standard) | 50-99-7 | 99.8% | 500 MG
SDP
Catalog No. 5000407286
Encompass_Preferred
D-Glucose (Standard) is the analytical standard for D-Glucose, intended for research and analytical applications. It is the naturally occurring form of glucose and the most abundant monosaccharide. D-Glucose plays a critical role in general metabolism and serves as a significant signaling molecule related to cellular metabolic status and stress responses.
  • Analytical standard for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for use in HPLC, GC, and MS.
Catalog No. 50-004-07286 Supplier Medchemexpress LLC Supplier No. HYB0389R500MG
D-Glucose (Standard) is the analytical standard for D-Glucose, intended for research and analytical applications. It is the naturally occurring form of glucose and the most abundant monosaccharide. D-Glucose plays a critical role in general metabolism and serves as a significant signaling molecule related to cellular metabolic status and stress responses.
  • Analytical standard for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for use in HPLC, GC, and MS.