Learn More
Medchemexpress LLC D-Glucosyl-β-1,1′-N-palmitoylsphingosine | 74365-77-8 | 700.04 | 1 MG
SDP
Catalog No. 5000414789
Encompass_Preferred
D-Glucosyl-β-1,1′-N-palmitoylsphingosine is an endogenous Mincle ligand with immunostimulatory activity. It binds to and activates macrophage-inducible C-type lectin (Mincle).
  • Applicable to the research of immune system diseases
  • Endogenous Mincle ligand
  • Immunostimulatory activity
Catalog No. 50-004-14789 Supplier Medchemexpress LLC Supplier No. HYN106341MG
D-Glucosyl-β-1,1′-N-palmitoylsphingosine is an endogenous Mincle ligand with immunostimulatory activity. It binds to and activates macrophage-inducible C-type lectin (Mincle).
  • Applicable to the research of immune system diseases
  • Endogenous Mincle ligand
  • Immunostimulatory activity