Learn More
Cayman Chemical D-Glucuronic Acid, Purity: ≥95%, CAS: 6556-12-3, 10 g
SDP
Catalog No. 502992059
Encompass_Preferred
A metabolite of glucose; a component of proteoglycans; levels are increased in fibroblasts isolated from patients with ISSD or Salla disease
Catalog No. 50-299-2059 Supplier Cayman Chemical Supplier No. 3153110 G
A metabolite of glucose; a component of proteoglycans; levels are increased in fibroblasts isolated from patients with ISSD or Salla disease