Learn More
Cayman Chemical D-Glucuronic Acid, Purity: ≥95%, CAS: 6556-12-3, 50 g
SDP
Catalog No. 502991088
Encompass_Preferred
A metabolite of glucose; a component of proteoglycans; levels are increased in fibroblasts isolated from patients with ISSD or Salla disease
Catalog No. 50-299-1088 Supplier Cayman Chemical Supplier No. 3153150 G
A metabolite of glucose; a component of proteoglycans; levels are increased in fibroblasts isolated from patients with ISSD or Salla disease