Learn More
Medchemexpress LLC D-Glucuronic acid | 6556-12-3 | 1 G
SDP
Catalog No. 5000418200
Encompass_Preferred
D-Glucuronic acid is a key component of anti-inflammatory proteoglycans, promoting embryonic development and inhibiting cell aggregation. When metabolized into ethyl glucuronide, it activates Toll-like receptor 4 (TLR4), contributing to pain. This acid and its derivative, glucurono-lactone, act as liver detoxifiers for human health and show anti-tumor activity.
  • Key component of anti-inflammatory proteoglycans.
  • Promotes embryonic development.
  • Inhibits cell aggregation.
  • Acts as a liver detoxifier.
  • Derivatives exhibit anti-tumor activity.
  • Activates Toll-like receptor 4 (TLR4) when metabolized.
Catalog No. 50-004-18200 Supplier Medchemexpress LLC Supplier No. HYN66121G
D-Glucuronic acid is a key component of anti-inflammatory proteoglycans, promoting embryonic development and inhibiting cell aggregation. When metabolized into ethyl glucuronide, it activates Toll-like receptor 4 (TLR4), contributing to pain. This acid and its derivative, glucurono-lactone, act as liver detoxifiers for human health and show anti-tumor activity.
  • Key component of anti-inflammatory proteoglycans.
  • Promotes embryonic development.
  • Inhibits cell aggregation.
  • Acts as a liver detoxifier.
  • Derivatives exhibit anti-tumor activity.
  • Activates Toll-like receptor 4 (TLR4) when metabolized.