Learn More
Sigma Aldrich Fine Chemicals Biosciences D-Glucuronic acid 98 GC10G
Catalog No. 502897442
Encompass
Glucuronic acid is one of the component of xylan backbone a hemicellulose present in plant cell wall.
Catalog No. 50-289-7442 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G526910G
Glucuronic acid is one of the component of xylan backbone a hemicellulose present in plant cell wall.