Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucuronic acid | 99.9% | 240.09 g·mol⁻¹ | 13C6H11NaO8 | 10 MG
SDP

Catalog No. 5000289724
Encompass_Preferred

13C6-labeled D-glucuronic acid sodium salt monohydrate provided as a stable-isotope internal standard for metabolomics and biochemical research. The material is the sodium salt monohydrate of D-glucuronic acid, isotopically labeled on all six carbons (13C6), and supplied as a high-purity dry solid.

  • 13C6 isotopic labeling for tracer and internal standard use.
  • High purity (HPLC) - 99.9%.
  • Molecular weight 240.09 g·mol⁻¹ and formula 13C6H11NaO8.
  • Suitable for LC-MS and metabolomics workflows as a standard or tracer.
  • Supplied as a stable dry solid for controlled storage.

Catalog No. 50-002-89724 Supplier Medchemexpress LLC Supplier No. HYN6612S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

13C6-labeled D-glucuronic acid sodium salt monohydrate provided as a stable-isotope internal standard for metabolomics and biochemical research. The material is the sodium salt monohydrate of D-glucuronic acid, isotopically labeled on all six carbons (13C6), and supplied as a high-purity dry solid.

  • 13C6 isotopic labeling for tracer and internal standard use.
  • High purity (HPLC) - 99.9%.
  • Molecular weight 240.09 g·mol⁻¹ and formula 13C6H11NaO8.
  • Suitable for LC-MS and metabolomics workflows as a standard or tracer.
  • Supplied as a stable dry solid for controlled storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.